Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rhodamine 123, 99+%, pure

Catalog No. AC419095000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419095000 500 mg Glass bottle
AC419091000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC419095000 Supplier Thermo Scientific Chemicals Supplier No. 419095000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 62669-70-9 | C21H19ClN2O4 | 398.843 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62669-70-9
Molecular Formula C21H19ClN2O4
Molecular Weight (g/mol) 398.843
MDL Number MFCD00012664
InChI Key SEXIURXGNASJRB-UHFFFAOYSA-N
PubChem CID 16211830
IUPAC Name methyl 2-(3-amino-6-iminoxanthen-9-yl)benzoate;hydrate;hydrochloride
SMILES COC(=O)C1=CC=CC=C1C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N.O.Cl

Specifications

Assay Percent Range 99+%
Merck Index 15, 8306
Infrared Spectrum Authentic
Formula Weight 380.82
Grade Extra Pure
Packaging Glass bottle
UV_Vis 99.0% min.
Color Brown to Brown/Red or Green
Quantity 500 mg
Physical Form Powder
Chemical Name or Material Rhodamine 123, 99+%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 263-687-8
RTECSNumber ZE0883000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.