accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OO

CAS RN 62669-74-3

IUPAC Name: 7-(dimethylamino)-1H,2H,3H,4H-cyclopenta[c]chromen-4-one
Synonyms: 7-(dimethylamino)-1H,2H,3H-cyclopenta[c]chromen-4-one
Molecular Weight (g/mol): 229.28
Molecular Formula: C14H15NO2
InChi Key: RIUSGHALMCFISX-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C2C3=C(CCC3)C(=O)OC2=C1
Color 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)