Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 138, Laser Grade 99%, Thermo Scientific™

Catalog No. AC405565000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC405565000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC405565000 Supplier Thermo Scientific Chemicals Supplier No. 405565000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62669-74-3
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.279
InChI Key RIUSGHALMCFISX-UHFFFAOYSA-N
Synonym 7-Dimethylaminocyclopenta[c]coumarin
PubChem CID 100527
IUPAC Name 7-(dimethylamino)-2,3-dihydro-1H-cyclopenta[c]chromen-4-one
SMILES CN(C)C1=CC2=C(C=C1)C3=C(CCC3)C(=O)O2

Specifications

Fluorescence Max 447nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 365nm (in methanol)
Formula Weight 229.28
Percent Purity ≥98.5%
Molar Absorption Coefficient 22300L/(mole.cm) (lambda max.)
Suitability (for use as a laser dye) passes
Color Green/Yellow to Yellow
Quantity 500 mg
Physical Form Crystals or Flakes
Chemical Name or Material Coumarin 138, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.