accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62697-73-8

IUPAC Name: 2-amino-4-methanesulfinylbutanoic acid
Synonyms: methionine sulfoxide|S-oxide-methionine|uethionine, S-oxide|S-oxide uethionine
Molecular Weight (g/mol): 165.21
Molecular Formula: C5H11NO3S
InChi Key: QEFRNWWLZKMPFJ-UHFFFAOYNA-N
SMILES: CS(=O)CCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)