Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Methionine Sulfoxide 98.0+%, TCI America™
SDP

Catalog No. M01015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M01015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M01015G Supplier TCI America Supplier No. M01015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62697-73-8
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00002620
InChI Key QEFRNWWLZKMPFJ-UHFFFAOYNA-N
Synonym dl-methionine sulfoxide, methionine sulfoxide, 2-amino-4-methylsulfinyl butanoic acid, dl-methioninesulfoxide, methionine, s-oxide, uethionine, s-oxide, butanoic acid, 2-amino-4-methylsulfinyl, 2-amino-4-methylsulfinyl butyric acid, 2-amino-4-methanesulfinylbutanoic acid, butyric acid, 2-amino-4-methylsulfinyl
PubChem CID 847
ChEBI CHEBI:49033
IUPAC Name 2-amino-4-methanesulfinylbutanoic acid
SMILES CS(=O)CCC(N)C(O)=O

Specifications

Quantity 5 g
Formula Weight 165.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Methionine Sulfoxide
EINECSNumber (9)-1431
RTECSNumber ES7120000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.