accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62698-54-8

IUPAC Name: tetrasodium 2-[({5-[1-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Synonyms: tetrasodium 2-[({5-[1-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular Weight (g/mol): 724.54
Molecular Formula: C32H28N2Na4O12
InChi Key: KSMPPVYJZJBIES-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
Quantity 
  • (2)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)