Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresolphthalein complexone tetrasodium salt

Catalog No. AAA1794103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1794103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1794103 Supplier Thermo Scientific Chemicals Supplier No. A1794103

missing translation for 'validMainframeMsg'

CAS: 62698-54-8 | C32H28N2Na4O12 | 724.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62698-54-8
Molecular Formula C32H28N2Na4O12
Molecular Weight (g/mol) 724.54
MDL Number MFCD00066371
InChI Key KSMPPVYJZJBIES-UHFFFAOYSA-J
Synonym phthalein complexon sodium salt, o-cresolphthalein complexone tetrasodium salt, sodium 2,2',2,2'-3-oxo-1,3-dihydroisobenzofuran-1,1-diyl bis 2-hydroxy-3-methyl-5,1-phenylene bis methylene bis azanetriyl tetraacetate, phthaleincomplexonsodiumsalt, phthalein complexone sodium salt, glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, sodium salt 1:4, glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, tetrasodium salt, tetrasodium 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-methylbenzene-3,1-diyl methanediylnitrilo tetraacetate, tetrasodium 2-5-1-3-bis carboxylatomethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl carboxylatomethyl amino acetate, tetrasodium 2-5-1-3-bis 2-oxido-2-oxoethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl-2-oxido-2-oxoethyl amino acetate
PubChem CID 112916
IUPAC Name tetrasodium;2-[[5-[1-[3-[[bis(carboxylatomethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1

Specifications

Quantity 1 g
Formula Weight 724.54
Chemical Name or Material o-Cresolphthalein complexone tetrasodium salt
EINECSNumber 305-335-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.