CAS RN 62698-56-0
IUPAC Name:
tetrasodium 2-{[(7-{[bis(carboxylatomethyl)amino]methyl}-9-(2-carboxyphenyl)-6-hydroxy-3-oxo-3H-xanthen-2-yl)methyl](carboxylatomethyl)amino}acetate
Synonyms:
tetrasodium 2-{[(7-{[bis(carboxylatomethyl)amino]methyl}-9-(2-carboxyphenyl)-6-hydroxy-3-oxoxanthen-2-yl)methyl](carboxylatomethyl)amino}acetate
Molecular Weight (g/mol):
710.47
Molecular Formula:
C30H22N2Na4O13
InChi Key:
WCODCQAZATVMQI-UHFFFAOYSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].OC(=O)C1=CC=CC=C1C1=C2C=C(CN(CC([O-])=O)CC([O-])=O)C(=O)C=C2OC2=CC(O)=C(CN(CC([O-])=O)CC([O-])=O)C=C12
1
–
1
of
1
results
1
Fluorexon, indicator grade, Thermo Scientific™
CAS: 62698-56-0 Molecular Formula: C30H22Na4N2O13·xH2O Synonym: Calcein,Fluorexon,Fluorescein Complexon