Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorexon, indicator grade, Thermo Scientific™

Catalog No. AC152890250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152890250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC152890250 Supplier Thermo Scientific Chemicals Supplier No. 152890250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62698-56-0
Molecular Formula C30H22Na4N2O13·xH2O
Synonym Calcein, Fluorexon, Fluorescein Complexon

Specifications

Formula Weight 710.46
Quantity 25 g
Chemical Name or Material Fluorexon, indicator grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.