accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCl

CAS RN 627-42-9

IUPAC Name: 1-chloro-2-methoxyethane
Synonyms: ethane, 1-chloro-2-methoxy-
Molecular Weight (g/mol): 94.54
Molecular Formula: C3H7ClO
InChi Key: XTIGGAHUZJWQMD-UHFFFAOYSA-N
SMILES: COCCCl
Boiling Point 
  • (6)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (6)
  • (1)
  • (3)
  • (3)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)