accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 627-45-2

IUPAC Name: N-ethylformamide
Synonyms: N-formylethylamine
Molecular Weight (g/mol): 73.1
Molecular Formula: C3H7NO
InChi Key: KERBAAIBDHEFDD-UHFFFAOYSA-N
SMILES: CCNC=O
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results