Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Ethylformamide

Catalog No. 0395150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-951-50ML 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-951-50ML Supplier Sigma Aldrich Supplier No. 0395150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Ethylformamide

Specifications

CAS 627-45-2
Density 0.950 g/mL (at 20°C (literature))
Boiling Point 202°C to 204°C
Molecular Formula C3H7NO
Refractive Index n20/D 1.432
Linear Formula HCONHC2H5
MDL Number MFCD00042672
Quantity 100 g
Synonym N-Formylethylamine
Molecular Weight (g/mol) 73.09
Percent Purity ≥99.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.