accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 627-50-9

IUPAC Name: (ethylsulfanyl)ethene
Synonyms: (ethylsulfanyl)ethene
Molecular Weight (g/mol): 88.17
Molecular Formula: C4H8S
InChi Key: AFGACPRTZOCNIW-UHFFFAOYSA-N
SMILES: CCSC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results