Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Vinyl Sulfide 93.0+%, TCI America™
SDP

Catalog No. E03355ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03355ML 5 mL
E033525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03355ML Supplier TCI America Supplier No. E03355ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 627-50-9
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.168
MDL Number MFCD00015180
InChI Key AFGACPRTZOCNIW-UHFFFAOYSA-N
PubChem CID 12320
IUPAC Name ethenylsulfanylethane
SMILES CCSC=C

Specifications

Color Colorless
Boiling Point 91°C
Quantity 5 mL
UN Number 1993
Formula Weight 88.17
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Vinyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.