accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2CH3CH2

CAS RN 627-58-7

IUPAC Name: 2,5-dimethylhexa-1,5-diene
Synonyms: 2,5-dimethyl-1,5-hexadiene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: DSAYAFZWRDYBQY-UHFFFAOYSA-N
SMILES: CC(=C)CCC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results