Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-1,5-hexadiene 98.0+%, TCI America™
SDP

Catalog No. D073425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D073425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D073425ML Supplier TCI America Supplier No. D073425ML

Chemical Identifiers

CAS 627-58-7
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008602
InChI Key DSAYAFZWRDYBQY-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1,5-hexadiene, bimethallyl, dimethallyl, 1,5-hexadiene, 2,5-dimethyl, unii-phn34350em, 2,5-hexadiene, 2,5-diene, 1, 2,5-dimethyl, 2,5-dimethyl-hexa-1,5-diene, 1,5-hexadiene,2,5-dimethyl
PubChem CID 12322
IUPAC Name 2,5-dimethylhexa-1,5-diene
SMILES CC(=C)CCC(C)=C

Specifications

Color Colorless
Boiling Point 116°C
Quantity 25 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-1,5-hexadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.