accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 627-63-4

IUPAC Name: (2E)-but-2-enedioyl dichloride
Synonyms: fumaryl chloride
Molecular Weight (g/mol): 152.96
Molecular Formula: C4H2Cl2O2
InChi Key: ZLYYJUJDFKGVKB-OWOJBTEDSA-N
SMILES: ClC(=O)\C=C\C(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (5)
  • (2)
  • (7)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)