Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumaryl chloride, 95%

Catalog No. AC165975000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165975000 500 mL Glass bottle
AC165970250 25 mL Glass bottle
AC165971000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC165975000 Supplier Thermo Scientific Chemicals Supplier No. 165975000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 627-63-4 | C4H2Cl2O2 | 152.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-63-4
Molecular Formula C4H2Cl2O2
Molecular Weight (g/mol) 152.96
MDL Number MFCD00000733
InChI Key ZLYYJUJDFKGVKB-OWOJBTEDSA-N
Synonym fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride
PubChem CID 5325504
IUPAC Name (E)-but-2-enedioyl dichloride
SMILES C(=CC(=O)Cl)C(=O)Cl

Specifications

Melting Point -2.0°C
Color Yellow
Density 1.4100g/mL
Boiling Point 161.0°C to 164.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4980 to 1.5010
Linear Formula ClCOCH=CHCOCl
Quantity 500 mL
Beilstein 02, 743
Specific Gravity 1.41
Solubility Information Solubility in water: decomposes
Formula Weight 152.96
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Fumaryl chloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-005-4
RTECSNumber LT2800000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.