accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 627-90-7

IUPAC Name: ethyl undecanoate
Synonyms: ethyl undecanoate
Molecular Weight (g/mol): 214.35
Molecular Formula: C13H26O2
InChi Key: IAFQYUQIAOWKSB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(=O)OCC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)