Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Undecanoate 98.0+%, TCI America™
SDP

Catalog No. U004925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004925ML 25 mL
U0049250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U004925ML Supplier TCI America Supplier No. U004925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 627-90-7
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.35
MDL Number MFCD00008958
InChI Key IAFQYUQIAOWKSB-UHFFFAOYSA-N
Synonym undecanoic acid, ethyl ester, ethyl undecylate, undecanoic acid ethyl ester, ethyl hendecanoate, ethyl n-undecanoate, n-undecanoic acid ethyl ester, fema no. 3492, ethyl undecanoate, acmc-1ayp4, undecanoic acid, ethylester
PubChem CID 12327
IUPAC Name ethyl undecanoate
SMILES CCCCCCCCCCC(=O)OCC

Specifications

Boiling Point 140°C
Quantity 25 mL
Formula Weight 214.35
Percent Purity ≥98.0% (GC)
Chemical Name or Material Ethyl Undecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.