accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6270-07-1

IUPAC Name: N-[2-(4-nitrophenyl)ethyl]acetamide
Synonyms: N-[2-(4-nitrophenyl)ethyl]acetamide
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: DIRUSBMKFDPKDI-UHFFFAOYSA-N
SMILES: CC(=O)NCCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results