Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-2-(4-nitrophenyl)ethylamine 98.0+%, TCI America™
SDP

Catalog No. A14945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14945G 5 g
A149425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14945G Supplier TCI America Supplier No. A14945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6270-07-1
Molecular Formula C10H12N2O3
Molecular Weight (g/mol) 208.22
MDL Number MFCD00024805
InChI Key DIRUSBMKFDPKDI-UHFFFAOYSA-N
Synonym N-Acetyl-4-nitrophenethylamine, N-[2-(4-Nitrophenyl)ethyl]acetamide
PubChem CID 80450
IUPAC Name N-[2-(4-nitrophenyl)ethyl]acetamide
SMILES CC(=O)NCCC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 140°C
Quantity 5 g
Formula Weight 208.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-2-(4-nitrophenyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.