accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 6271-23-4

IUPAC Name: ethyl 11-bromoundecanoate
Synonyms: ethyl 11-bromoundecanoate
Molecular Weight (g/mol): 293.25
Molecular Formula: C13H25BrO2
InChi Key: RGWOAXNKJWTDFA-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCCCCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results