Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 11-Bromoundecanoate 96.0+%, TCI America™
SDP

Catalog No. B22425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22425G Supplier TCI America Supplier No. B22425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6271-23-4
Molecular Formula C13H25BrO2
Molecular Weight (g/mol) 293.25
MDL Number MFCD00045048
InChI Key RGWOAXNKJWTDFA-UHFFFAOYSA-N
Synonym 11-Bromoundecanoic Acid Ethyl Ester
PubChem CID 235144
IUPAC Name ethyl 11-bromoundecanoate
SMILES CCOC(=O)CCCCCCCCCCBr

Specifications

Color Yellow
Quantity 5 g
Formula Weight 293.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 11-Bromoundecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.