accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6271-79-0

IUPAC Name: 3,3'-dinitro-[1,1'-biphenyl]-4,4'-diamine
Synonyms: 3,3'-dinitro-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 274.24
Molecular Formula: C12H10N4O4
InChi Key: OCEINMLGYDSKFW-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1[N+]([O-])=O)C1=CC=C(N)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results