Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Dinitrobenzidine 95.0+%, TCI America™
SDP

Catalog No. D082225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D082225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D082225G Supplier TCI America Supplier No. D082225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6271-79-0
Molecular Formula C12H10N4O4
Molecular Weight (g/mol) 274.236
MDL Number MFCD00024264
InChI Key OCEINMLGYDSKFW-UHFFFAOYSA-N
Synonym 4,4′C-Diamino-3,3′C-dinitrobiphenyl
PubChem CID 3106720
IUPAC Name 4-(4-amino-3-nitrophenyl)-2-nitroaniline
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N)[N+](=O)[O-])[N+](=O)[O-])N

Specifications

Melting Point 275°C
Quantity 25 g
Formula Weight 274.24
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dinitrobenzidine
RTECSNumber DD1250000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.