accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSClCl

CAS RN 6274-27-7

IUPAC Name: 2-[(2,5-dichlorophenyl)sulfanyl]acetic acid
Synonyms: [(2,5-dichlorophenyl)sulfanyl]acetic acid
Molecular Weight (g/mol): 237.09
Molecular Formula: C8H6Cl2O2S
InChi Key: DCXXCTCEJFBLPD-UHFFFAOYSA-N
SMILES: OC(=O)CSC1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results