accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNNCl

CAS RN 6275-01-0

IUPAC Name: 5-methyl-2,3-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 5-methyl-2,3-diphenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 272.74
Molecular Formula: C14H13ClN4
InChi Key: DNLGEYNSADNPQS-UHFFFAOYSA-M
SMILES: [Cl-].CC1=NN(C2=CC=CC=C2)[N+](=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)