Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenyl-5-methyltetrazolium Chloride 95.0+%, TCI America™
SDP

Catalog No. B0313100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0313100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0313100MG Supplier TCI America Supplier No. B0313100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6275-01-0
Molecular Formula C14H13ClN4
Molecular Weight (g/mol) 272.74
MDL Number MFCD00060025
InChI Key DNLGEYNSADNPQS-UHFFFAOYSA-M

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 272.74
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenyl-5-methyltetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.