CAS RN 627518-40-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PDGFR Tyrosine Kinase Inhibitor IV
A cell-permeable indenopyrazole compound that displays potent antiproliferative properties in several human tumor cell lines (IC50 < 33
2
R&D Systems™ JNJ 10198409
A Tocris product, part of R&D Systems. Potent PDGFRα and PDGFRβ inhbitor; also inhibits c-Abl, Lck, c-Src and Fyn kinases