accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CClF

CAS RN 627525-83-1

IUPAC Name: 2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 256.51
Molecular Formula: C12H15BClFO2
InChi Key: XJZYKURDYQAGGN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results