accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3NCH3NNSOHOO(E/Z)

CAS RN 62758-12-7

IUPAC Name: 4-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzene-1-sulfonic acid sodium
Synonyms: 4-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzenesulfonic acid sodium
Molecular Weight (g/mol): 356.4
Molecular Formula: C16H19N3NaO3S
InChi Key: KEFGAXCAAQPJJK-UHFFFAOYSA-N
SMILES: [Na].CCN(CC)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)