Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Orange sodium salt, Thermo Scientific Chemicals

Catalog No. AAJ6286322
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6286322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6286322 Supplier Thermo Scientific Chemicals Supplier No. J6286322
Add to Cart

missing translation for 'validMainframeMsg'

Indicator: pH 3.0 (red) to pH 4.5 (yellow)

Indicator: pH 3.0 (red) to pH 4.5 (yellow)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62758-12-7
Molecular Formula C16H18N3NaO3S
Molecular Weight (g/mol) 355.388
MDL Number MFCD00007503
InChI Key YZORUOZKRBVLEG-UHFFFAOYSA-M
Synonym 4-[4-(Diethylamino)phenylazo]benzenesulfonic acid sodium salt
PubChem CID 23676309
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Beilstein 3821641
Color Orange
Quantity 100 g
Physical Form Powder
Formula Weight 355.39
Chemical Name or Material Ethyl Orange sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 263-716-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.