accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-28-4

IUPAC Name: 1-methoxybutane
Synonyms: methyl N-butyl ether
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: CXBDYQVECUFKRK-UHFFFAOYSA-N
SMILES: CCCCOC
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)