accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 628-55-7

IUPAC Name: 2-methyl-1-(2-methylpropoxy)propane
Synonyms: di-isobutyl ether
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: SZNYYWIUQFZLLT-UHFFFAOYSA-N
SMILES: CC(C)COCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results