accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 628-73-9

IUPAC Name: hexanenitrile
Synonyms: hexanenitrile
Molecular Weight (g/mol): 97.16
Molecular Formula: C6H11N
InChi Key: AILKHAQXUAOOFU-UHFFFAOYSA-N
SMILES: CCCCCC#N
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)