Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanenitrile 97.0+%, TCI America™
SDP

Catalog No. H010225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H010225ML 25 mL
H01025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H010225ML Supplier TCI America Supplier No. H010225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 628-73-9
Molecular Formula C6H11N
Molecular Weight (g/mol) 97.161
MDL Number MFCD00001979
InChI Key AILKHAQXUAOOFU-UHFFFAOYSA-N
Synonym Amyl Cyanide, Capronitrile
PubChem CID 12352
IUPAC Name hexanenitrile
SMILES CCCCCC#N

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
UN Number 1993
Formula Weight 97.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexanenitrile
RTECSNumber MO3900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.