accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH

CAS RN 62803-47-8

IUPAC Name: 6-hydroxy-2,3-dihydro-1H-inden-1-one
Synonyms: 6-hydroxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: MOANRQDXNNXOLW-UHFFFAOYSA-N
SMILES: OC1=CC2=C(CCC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results