Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-1-indanone 98.0+%, TCI America™
SDP

Catalog No. H13105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13105G 5 g
H13101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13105G Supplier TCI America Supplier No. H13105G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62803-47-8
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00513530
InChI Key MOANRQDXNNXOLW-UHFFFAOYSA-N
PubChem CID 7020659
IUPAC Name 6-hydroxy-2,3-dihydro-1H-inden-1-one
SMILES OC1=CC2=C(CCC2=O)C=C1

Specifications

Melting Point 157°C
Color Yellow
Quantity 5 g
Formula Weight 148.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.