accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6284-83-9

IUPAC Name: 1,3,5-trichloro-2,4-dinitrobenzene
Synonyms: benzene, dinitrotrichloro-
Molecular Weight (g/mol): 271.43
Molecular Formula: C6HCl3N2O4
InChi Key: BPMOJGOPWSCNHJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C(=C(Cl)C=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results