Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dinitro-2,4,5-trichlorobenzene, MP Biomedicals

Catalog No. ICN205760 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205760 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205760 Supplier MP Biomedicals, Inc Supplier No. 0520576001

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6284-83-9
Molecular Formula C6HCl3N2O4
Molecular Weight (g/mol) 271.434
InChI Key BPMOJGOPWSCNHJ-UHFFFAOYSA-N
Synonym benzene, dinitrotrichloro, dinitrotrichlorobenzene, 1,3-dinitro-2,4,6-trichlorobenzene, 2,4-dinitro-1,3,5-trichlorobenzene, benzene, 1,3,5-trichloro-2,4-dinitro, 2,4,6-trichloro-1,3-dinitrobenzene, trichloro-1,3-dinitrobenzene, benzene, trichloro-1,3-dinitro, benzene, 2,4-dinitro-1,3,5-trichloro, ambsclk-291
PubChem CID 22696
IUPAC Name 1,3,5-trichloro-2,4-dinitrobenzene
SMILES C1=C(C(=C(C(=C1Cl)[N+](=O)[O-])Cl)[N+](=O)[O-])Cl

Specifications

Quantity 10 g
Formula Weight 271.44
Chemical Name or Material 1,3-Dinitro-2,4,5-Trichlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.