CAS RN 62846-82-6
IUPAC Name:
ethyl pyrimidine-4-carboxylate
Synonyms:
ethyl pyrimidine-4-carboxylate
Molecular Weight (g/mol):
152.15
Molecular Formula:
C7H8N2O2
InChi Key:
DWRWSNAREGLUHZ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NC=NC=C1