accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSNNOO

CAS RN 6285-57-0

IUPAC Name: 6-nitro-1,3-benzothiazol-2-amine
Synonyms: 6-nitro-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 195.2
Molecular Formula: C7H5N3O2S
InChi Key: GPNAVOJCQIEKQF-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=C(C=C2S1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results