CAS RN 62855-02-1
IUPAC Name:
(1,2,3,4-tetrahydroisoquinolin-3-yl)methanol
Synonyms:
1,2,3,4-tetrahydroisoquinolin-3-ylmethanol
Molecular Weight (g/mol):
163.22
Molecular Formula:
C10H13NO
InChi Key:
ZSKDXMLMMQFHGW-UHFFFAOYNA-N
SMILES:
OCC1CC2=CC=CC=C2CN1
1
–
1
of
1
results
1
(R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol, 97%, Thermo Scientific™
CAS: 62855-02-1 Molecular Formula: C10H13NO Molecular Weight (g/mol): 163.22 MDL Number: MFCD02684327 InChI Key: ZSKDXMLMMQFHGW-SNVBAGLBSA-N Synonym: r-1,2,3,4-tetrahydroisoquinolin-3-yl methanol,3r-1,2,3,4-tetrahydroisoquinolin-3-ylmethanol,r-1,2,3,4-tetrahydroisoquinolin-3-yl-methanol,r-1,2,3,4-tetrahydroisoquinolylmethan-3-ol,3-isoquinolinemethanol, 1,2,3,4-tetrahydro-, r,chembl61490,3r-1,2,3,4-tetrahydroisoquinolin-3-yl methanol,r-3-hydroxymethyl-1,2,3,4-tetrahydroisoquinoline,ksc288q9p PubChem CID: 723422 IUPAC Name: [(3R)-1,2,3,4-tetrahydroisoquinolin-3-yl]methanol SMILES: C1C(NCC2=CC=CC=C21)CO