CAS RN 6286-30-2
CAS RN 6286-30-2
IUPAC Name:
2,3-dibromo-3-phenylpropanoic acid
Synonyms:
2,3-dibromo-3-phenylpropanoic acid
Molecular Weight (g/mol):
307.97
Molecular Formula:
C9H8Br2O2
InChi Key:
FXJWTHBNVZNQQP-UHFFFAOYNA-N
SMILES:
OC(=O)C(Br)C(Br)C1=CC=CC=C1
1
–
1
of
1
results
1
2,3-Dibromo-3-phenylpropionic Acid 98.0+%, TCI America™

CAS: 6286-30-2 Molecular Formula: C9H8Br2O2 Molecular Weight (g/mol): 307.97 MDL Number: MFCD00004208 InChI Key: FXJWTHBNVZNQQP-UHFFFAOYNA-N Synonym: alpha,beta-Dibromohydrocinnamic Acid PubChem CID: 22705 IUPAC Name: 2,3-dibromo-3-phenylpropanoic acid SMILES: OC(=O)C(Br)C(Br)C1=CC=CC=C1