accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOHO

CAS RN 6286-43-7

IUPAC Name: cyclohexane-1,2,3-triol
Synonyms: cyclohexane-1,2,3-triol
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: IZSANPWSFUSNMY-UHFFFAOYNA-N
SMILES: OC1CCCC(O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results