accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBrOCH3OHO

CAS RN 6286-46-0

IUPAC Name: 2-bromo-4,5-dimethoxybenzoic acid
Synonyms: 2-bromo-4,5-dimethoxybenzoic acid
Molecular Weight (g/mol): 261.07
Molecular Formula: C9H9BrO4
InChi Key: HWFCHCRFQWEFMU-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(C=C1OC)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results