accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 629-14-1

IUPAC Name: 1,2-diethoxyethane
Synonyms: 1,2-diethoxyethane
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: LZDKZFUFMNSQCJ-UHFFFAOYSA-N
SMILES: CCOCCOCC
Molecular Weight (g/mol) 
  • (5)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)