accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 629-23-2

IUPAC Name: tetradecan-3-one
Synonyms: 3-tetradecanone
Molecular Weight (g/mol): 212.38
Molecular Formula: C14H28O
InChi Key: OCHYRSKMMMYUMI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results