Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Tetradecanone 95.0+%, TCI America™
SDP

Catalog No. T17601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17601G Supplier TCI America Supplier No. T17601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-23-2
Molecular Formula C14H28O
Molecular Weight (g/mol) 212.377
MDL Number MFCD00026563
InChI Key OCHYRSKMMMYUMI-UHFFFAOYSA-N
Synonym Ethyl Undecyl Ketone
PubChem CID 69418
IUPAC Name tetradecan-3-one
SMILES CCCCCCCCCCCC(=O)CC

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 152°C
Quantity 1 g
Formula Weight 212.38
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Tetradecanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.